Assay Design



Gene Transcripts

Gene Symbol
TVP23C
Entrez Gene ID
201158
Gene Name
trans-golgi network vesicle protein 23 homolog C (S. cerevisiae)
Gene Aliases
FAM18B2
Chromosome Location
Chr.17: 15502264 - 15563631 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145301.2 NP_660344.2 6 - 6 69
GenBank mRNA BC011952.1 - 6 - 6 69
BQ448398.1 - 1 - 1 232 69

View Full Product Details