Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128608.1 | NP_001122080.1 | 19 - 20 | 2396 | 87 |
NM_001265611.1 | NP_001252540.1 | 18 - 19 | 2378 | 87 | |
NM_014994.2 | NP_055809.2 | 18 - 19 | 2378 | 87 | |
NR_049761.1 | 17 - 18 | 2188 | 87 | ||
NR_049762.1 | 16 - 17 | 2139 | 87 | ||
XM_006720438.1 | XP_006720501.1 | 17 - 18 | 2195 | 87 | |
XM_006720439.2 | XP_006720502.1 | 5 - 6 | 470 | 87 | |
XM_011521383.1 | XP_011519685.1 | 18 - 19 | 2213 | 87 | |
XM_011521384.2 | XP_011519686.1 | 19 - 20 | 2360 | 87 | |
XM_017022017.1 | XP_016877506.1 | 17 - 18 | 2489 | 87 | |
GenBank mRNA | AB011168.2 | - | 19 - 20 | 2306 | 87 |
AB177850.1 | - | 10 - 11 | 1226 | 87 | |
AK302482.1 | - | 16 - 17 | 2080 | 87 | |
AL833267.1 | - | 18 - 19 | 2298 | 87 | |
BC036660.1 | - | 17 - 18 | 2193 | 87 | |
BC113983.1 | - | 18 - 19 | 2270 | 87 | |
BC114493.1 | - | 19 - 20 | 2288 | 87 | |
BX648786.1 | - | 18 - 19 | 2329 | 87 |