Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128608.1 | NP_001122080.1 | 21 - 22 | 2589 | 70 |
NM_001265611.1 | NP_001252540.1 | 20 - 21 | 2571 | 70 | |
NM_014994.2 | NP_055809.2 | 20 - 21 | 2571 | 70 | |
NR_049761.1 | 19 - 20 | 2381 | 70 | ||
NR_049762.1 | 18 - 19 | 2332 | 70 | ||
XM_006720438.1 | XP_006720501.1 | 19 - 20 | 2388 | 70 | |
XM_006720439.2 | XP_006720502.1 | 7 - 8 | 663 | 70 | |
XM_011521383.1 | XP_011519685.1 | 20 - 21 | 2406 | 70 | |
XM_011521384.2 | XP_011519686.1 | 21 - 22 | 2553 | 70 | |
XM_017022017.1 | XP_016877506.1 | 19 - 20 | 2682 | 70 | |
GenBank mRNA | AB011168.2 | - | 21 - 22 | 2499 | 70 |
AB177850.1 | - | 12 - 13 | 1419 | 70 | |
AK302482.1 | - | 18 - 19 | 2273 | 70 | |
AL833267.1 | - | 20 - 21 | 2491 | 70 | |
BC036660.1 | - | 19 - 20 | 2386 | 70 | |
BC113983.1 | - | 20 - 21 | 2463 | 70 | |
BC114493.1 | - | 21 - 22 | 2481 | 70 | |
BX640787.1 | - | 17 - 18 | 2736 | 70 | |
BX648786.1 | - | 20 - 21 | 2522 | 70 |