Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128608.1 | NP_001122080.1 | 22 - 23 | 2733 | 95 |
NM_001265611.1 | NP_001252540.1 | 21 - 22 | 2715 | 95 | |
NM_014994.2 | NP_055809.2 | 21 - 22 | 2715 | 95 | |
NR_049761.1 | 20 - 21 | 2525 | 95 | ||
NR_049762.1 | 19 - 20 | 2476 | 95 | ||
XM_006720438.1 | XP_006720501.1 | 20 - 21 | 2532 | 95 | |
XM_006720439.2 | XP_006720502.1 | 8 - 9 | 807 | 95 | |
XM_011521383.1 | XP_011519685.1 | 21 - 22 | 2550 | 95 | |
XM_011521384.2 | XP_011519686.1 | 22 - 23 | 2697 | 95 | |
XM_017022017.1 | XP_016877506.1 | 20 - 21 | 2826 | 95 | |
GenBank mRNA | AB011168.2 | - | 22 - 23 | 2643 | 95 |
AB177850.1 | - | 13 - 14 | 1563 | 95 | |
AK302482.1 | - | 19 - 20 | 2417 | 95 | |
AL833267.1 | - | 21 - 22 | 2635 | 95 | |
BC036660.1 | - | 20 - 21 | 2530 | 95 | |
BC113983.1 | - | 21 - 22 | 2607 | 95 | |
BC114493.1 | - | 22 - 23 | 2625 | 95 | |
BX648786.1 | - | 21 - 22 | 2666 | 95 |