Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173611.3 | NP_775882.2 | 3 - 4 | 378 | 155 |
GenBank mRNA | AK095745.1 | - | 3 - 4 | 352 | 155 |
BC035600.1 | - | 2 - 3 | 236 | 155 | |
BC093898.1 | - | 3 - 4 | 348 | 155 | |
BP872319.1 | - | 3 - 4 | 378 | 155 | |
BP872410.1 | - | 3 - 4 | 378 | 155 |