Assay Design



Gene Transcripts

Gene Symbol
FAM98B
Entrez Gene ID
283742
Gene Name
family with sequence similarity 98 member B
Gene Aliases
-
Chromosome Location
Chr.15: 38454127 - 38487710 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173611.3 NP_775882.2 4 - 5 567 118
GenBank mRNA AK095745.1 - 4 - 5 541 118
BC035600.1 - 3 - 4 425 118
BC093898.1 - 4 - 5 537 118

View Full Product Details