Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004598.3 | NP_004589.1 | 10 - 11 | 1284 | 68 |
GenBank mRNA | AB209701.1 | - | 11 - 12 | 1268 | 68 |
AF231124.1 | - | 10 - 11 | 1154 | 68 | |
AK094489.1 | - | 9 - 10 | 1324 | 68 | |
BC030691.2 | - | 10 - 11 | 1265 | 68 |