Assay Design



Gene Transcripts

Gene Symbol
SPOCK1
Entrez Gene ID
6695
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1
Gene Aliases
SPOCK, TESTICAN, TIC1
Chromosome Location
Chr.5: 136975298 - 137499329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004598.3 NP_004589.1 2 - 3 341 72
GenBank mRNA AF231124.1 - 2 - 3 211 72
AK094489.1 - 1 - 2 381 72
BC030691.2 - 2 - 3 322 72

View Full Product Details