Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021044.3 | NP_066382.1 | 1 - 2 | 606 | 81 |
GenBank mRNA | AK313360.1 | - | 1 - 2 | 576 | 81 |
BC033507.1 | - | 1 - 2 | 606 | 81 | |
HQ607677.1 | - | 1 - 2 | 603 | 78 | |
HQ607678.1 | - | 1 - 2 | 610 | 81 |