Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033449.2 | NP_258260.1 | 5 - 6 | 422 | 56 |
XM_005268524.4 | XP_005268581.1 | 5 - 6 | 449 | 56 | |
XM_006714803.3 | XP_006714866.1 | 5 - 6 | 455 | 56 | |
XM_011537698.2 | XP_011536000.1 | 5 - 6 | 455 | 56 | |
XM_011537700.2 | XP_011536002.1 | 5 - 6 | 456 | 56 | |
XM_011537701.2 | XP_011536003.1 | 5 - 6 | 456 | 56 | |
XM_017010013.1 | XP_016865502.1 | 5 - 6 | 455 | 56 | |
GenBank mRNA | AK000007.1 | - | 5 - 6 | 406 | 56 |
AY217346.1 | - | 5 - 6 | 422 | 56 | |
BC047016.1 | - | 4 - 5 | 349 | 56 |