Assay Design



Gene Transcripts

Gene Symbol
FCHSD1
Entrez Gene ID
89848
Gene Name
FCH and double SH3 domains 1
Gene Aliases
NWK2
Chromosome Location
Chr.5: 141639302 - 141651453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033449.2 NP_258260.1 7 - 8 623 62
XM_005268524.4 XP_005268581.1 7 - 8 650 62
XM_011537698.2 XP_011536000.1 7 - 8 656 62
XM_011537700.2 XP_011536002.1 7 - 8 657 62
XM_011537701.2 XP_011536003.1 7 - 8 657 62
XM_017010013.1 XP_016865502.1 7 - 8 656 62
GenBank mRNA AK000007.1 - 7 - 8 607 62
AY217346.1 - 7 - 8 623 62
BC047016.1 - 6 - 7 550 62

View Full Product Details