Assay Design



Gene Transcripts

Gene Symbol
FCHSD1
Entrez Gene ID
89848
Gene Name
FCH and double SH3 domains 1
Gene Aliases
NWK2
Chromosome Location
Chr.5: 141639302 - 141651453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033449.2 NP_258260.1 9 - 10 878 97
XM_005268524.4 XP_005268581.1 9 - 10 905 97
XM_006714803.3 XP_006714866.1 8 - 9 782 97
XM_011537698.2 XP_011536000.1 9 - 10 911 97
XM_011537700.2 XP_011536002.1 9 - 10 912 97
XM_011537701.2 XP_011536003.1 9 - 10 912 97
XM_017010013.1 XP_016865502.1 9 - 10 911 97
GenBank mRNA AK000007.1 - 9 - 10 862 97
AY217346.1 - 9 - 10 878 97
AY358478.1 - 1 - 2 85 97
BC047016.1 - 8 - 9 805 97

View Full Product Details