Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142673.2 | NP_001136145.1 | 11 - 12 | 1468 | 117 |
NM_001205120.1 | NP_001192049.1 | 10 - 11 | 1412 | 117 | |
XM_011520501.1 | XP_011518803.1 | 12 - 13 | 1555 | 117 | |
XM_017018602.1 | XP_016874091.1 | 12 - 13 | 1705 | 117 | |
XM_017018603.1 | XP_016874092.1 | 13 - 14 | 1822 | 117 | |
XM_017018604.1 | XP_016874093.1 | 11 - 12 | 1499 | 117 | |
XM_017018605.1 | XP_016874094.1 | 12 - 13 | 1490 | 117 | |
XM_017018606.1 | XP_016874095.1 | 11 - 12 | 1434 | 117 | |
XM_017018607.1 | XP_016874096.1 | 9 - 10 | 811 | 117 | |
XM_017018619.1 | XP_016874108.1 | 11 - 12 | 1578 | 117 | |
XM_017018620.1 | XP_016874109.1 | 11 - 12 | 1638 | 117 | |
XM_017018621.1 | XP_016874110.1 | 10 - 11 | 1522 | 117 | |
GenBank mRNA | AB014552.1 | - | 10 - 11 | 1092 | 117 |
AK289641.1 | - | 11 - 12 | 1015 | 117 |