Assay Design



Gene Transcripts

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
Chromosome Location
Chr.11: 46617276 - 46676019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142673.2 NP_001136145.1 14 - 15 1836 113
NM_001205119.1 NP_001192048.1 14 - 15 1879 113
NM_001205120.1 NP_001192049.1 13 - 14 1780 113
NM_001205121.1 NP_001192050.1 13 - 14 1720 113
NM_001205122.1 NP_001192051.1 11 - 12 1587 113
NM_014741.4 NP_055556.2 13 - 14 1725 113
XM_005253262.2 XP_005253319.1 15 - 16 1675 113
XM_005253263.2 XP_005253320.1 15 - 16 1966 113
XM_005253265.2 XP_005253322.1 16 - 17 2172 113
XM_005253266.2 XP_005253323.1 15 - 16 1901 113
XM_005253268.2 XP_005253325.1 13 - 14 1278 113
XM_006718394.2 XP_006718457.1 16 - 17 2022 113
XM_006718395.2 XP_006718458.1 16 - 17 1957 113
XM_006718396.2 XP_006718459.1 15 - 16 2056 113
XM_011520493.1 XP_011518795.1 17 - 18 2159 113
XM_011520496.2 XP_011518798.1 14 - 15 1934 113
XM_011520499.2 XP_011518801.1 14 - 15 1548 113
XM_011520500.1 XP_011518802.1 16 - 17 1995 113
XM_011520501.1 XP_011518803.1 15 - 16 1923 113
XM_011520502.1 XP_011518804.1 15 - 16 1896 113
XM_011520503.1 XP_011518805.1 14 - 15 1812 113
XM_017018599.1 XP_016874088.1 16 - 17 2107 113
XM_017018600.1 XP_016874089.1 15 - 16 1648 113
XM_017018601.1 XP_016874090.1 14 - 15 1823 113
XM_017018602.1 XP_016874091.1 15 - 16 2073 113
XM_017018603.1 XP_016874092.1 16 - 17 2190 113
XM_017018604.1 XP_016874093.1 14 - 15 1867 113
XM_017018605.1 XP_016874094.1 15 - 16 1858 113
XM_017018606.1 XP_016874095.1 14 - 15 1802 113
XM_017018607.1 XP_016874096.1 12 - 13 1179 113
XM_017018608.1 XP_016874097.1 14 - 15 1549 113
XM_017018609.1 XP_016874098.1 14 - 15 1840 113
XM_017018610.1 XP_016874099.1 13 - 14 1724 113
XM_017018611.1 XP_016874100.1 14 - 15 1775 113
XM_017018612.1 XP_016874101.1 14 - 15 1962 113
XM_017018613.1 XP_016874102.1 13 - 14 1756 113
XM_017018614.1 XP_016874103.1 14 - 15 1747 113
XM_017018615.1 XP_016874104.1 13 - 14 1846 113
XM_017018616.1 XP_016874105.1 12 - 13 1640 113
XM_017018617.1 XP_016874106.1 14 - 15 1845 113
XM_017018618.1 XP_016874107.1 16 - 17 2161 113
XM_017018619.1 XP_016874108.1 14 - 15 1946 113
XM_017018620.1 XP_016874109.1 14 - 15 2006 113
XM_017018621.1 XP_016874110.1 13 - 14 1890 113
XM_017018622.1 XP_016874111.1 13 - 14 1847 113
GenBank mRNA AB014552.1 - 13 - 14 1460 113
AK023867.1 - 13 - 14 1458 113
AK223603.1 - 14 - 15 1555 113
AK289641.1 - 14 - 15 1383 113
AK294110.1 - 11 - 12 1300 113
AK296526.1 - 6 - 7 667 113
AK315925.1 - 6 - 7 594 113
BC001331.1 - 13 - 14 1154 113
BC002378.2 - 13 - 14 1424 113
BC006191.2 - 13 - 14 1466 113

View Full Product Details