Assay Design



Gene Transcripts

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
Chromosome Location
Chr.11: 46617276 - 46676019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142673.2 NP_001136145.1 6 - 7 992 145
NM_001205119.1 NP_001192048.1 5 - 6 936 145
NM_001205120.1 NP_001192049.1 5 - 6 936 145
NM_001205121.1 NP_001192050.1 6 - 7 987 145
NM_001205122.1 NP_001192051.1 4 - 5 854 145
NM_014741.4 NP_055556.2 6 - 7 992 145
XM_005253262.2 XP_005253319.1 6 - 7 732 145
XM_005253263.2 XP_005253320.1 6 - 7 1023 145
XM_005253265.2 XP_005253322.1 7 - 8 1229 145
XM_005253266.2 XP_005253323.1 6 - 7 958 145
XM_005253268.2 XP_005253325.1 4 - 5 335 145
XM_006718394.2 XP_006718457.1 7 - 8 1079 145
XM_006718395.2 XP_006718458.1 7 - 8 1014 145
XM_006718396.2 XP_006718459.1 6 - 7 1113 145
XM_011520493.1 XP_011518795.1 8 - 9 1216 145
XM_011520496.2 XP_011518798.1 5 - 6 991 145
XM_011520499.2 XP_011518801.1 5 - 6 605 145
XM_011520500.1 XP_011518802.1 7 - 8 1079 145
XM_011520501.1 XP_011518803.1 7 - 8 1079 145
XM_011520502.1 XP_011518804.1 7 - 8 1079 145
XM_011520503.1 XP_011518805.1 7 - 8 1079 145
XM_017018599.1 XP_016874088.1 7 - 8 1164 145
XM_017018600.1 XP_016874089.1 6 - 7 732 145
XM_017018601.1 XP_016874090.1 5 - 6 907 145
XM_017018602.1 XP_016874091.1 7 - 8 1229 145
XM_017018603.1 XP_016874092.1 8 - 9 1346 145
XM_017018604.1 XP_016874093.1 6 - 7 1023 145
XM_017018605.1 XP_016874094.1 7 - 8 1014 145
XM_017018606.1 XP_016874095.1 6 - 7 958 145
XM_017018607.1 XP_016874096.1 4 - 5 335 145
XM_017018608.1 XP_016874097.1 6 - 7 732 145
XM_017018609.1 XP_016874098.1 6 - 7 1023 145
XM_017018610.1 XP_016874099.1 5 - 6 907 145
XM_017018611.1 XP_016874100.1 6 - 7 958 145
XM_017018612.1 XP_016874101.1 7 - 8 1229 145
XM_017018613.1 XP_016874102.1 6 - 7 1023 145
XM_017018614.1 XP_016874103.1 7 - 8 1014 145
XM_017018615.1 XP_016874104.1 6 - 7 1113 145
XM_017018616.1 XP_016874105.1 5 - 6 907 145
XM_017018617.1 XP_016874106.1 7 - 8 1112 145
XM_017018618.1 XP_016874107.1 7 - 8 1079 145
XM_017018619.1 XP_016874108.1 6 - 7 963 145
XM_017018620.1 XP_016874109.1 6 - 7 1023 145
XM_017018621.1 XP_016874110.1 5 - 6 907 145
XM_017018622.1 XP_016874111.1 6 - 7 975 145
GenBank mRNA AB014552.1 - 5 - 6 616 145
AK023867.1 - 6 - 7 725 145
AK223603.1 - 5 - 6 612 145
AK289641.1 - 6 - 7 539 145
AK293109.1 - 6 - 7 691 145
AK294110.1 - 4 - 5 567 145
BC001331.1 - 6 - 7 421 145
BC002378.2 - 6 - 7 691 145
BC006191.2 - 6 - 7 733 145
CD675643.1 - 4 - 5 311 145

View Full Product Details