Assay Design



Gene Transcripts

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
Chromosome Location
Chr.11: 46617276 - 46676019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142673.2 NP_001136145.1 9 - 10 1279 113
NM_001205119.1 NP_001192048.1 8 - 9 1223 113
NM_001205120.1 NP_001192049.1 8 - 9 1223 113
NM_001205121.1 NP_001192050.1 9 - 10 1274 113
NM_001205122.1 NP_001192051.1 7 - 8 1141 113
NM_014741.4 NP_055556.2 9 - 10 1279 113
XM_005253262.2 XP_005253319.1 9 - 10 1019 113
XM_005253263.2 XP_005253320.1 9 - 10 1310 113
XM_005253265.2 XP_005253322.1 10 - 11 1516 113
XM_005253266.2 XP_005253323.1 9 - 10 1245 113
XM_005253268.2 XP_005253325.1 7 - 8 622 113
XM_006718394.2 XP_006718457.1 10 - 11 1366 113
XM_006718395.2 XP_006718458.1 10 - 11 1301 113
XM_006718396.2 XP_006718459.1 9 - 10 1400 113
XM_011520493.1 XP_011518795.1 11 - 12 1503 113
XM_011520496.2 XP_011518798.1 8 - 9 1278 113
XM_011520499.2 XP_011518801.1 8 - 9 892 113
XM_011520500.1 XP_011518802.1 10 - 11 1366 113
XM_011520501.1 XP_011518803.1 10 - 11 1366 113
XM_011520502.1 XP_011518804.1 10 - 11 1366 113
XM_011520503.1 XP_011518805.1 10 - 11 1366 113
XM_017018599.1 XP_016874088.1 10 - 11 1451 113
XM_017018600.1 XP_016874089.1 9 - 10 1019 113
XM_017018601.1 XP_016874090.1 8 - 9 1194 113
XM_017018602.1 XP_016874091.1 10 - 11 1516 113
XM_017018603.1 XP_016874092.1 11 - 12 1633 113
XM_017018604.1 XP_016874093.1 9 - 10 1310 113
XM_017018605.1 XP_016874094.1 10 - 11 1301 113
XM_017018606.1 XP_016874095.1 9 - 10 1245 113
XM_017018607.1 XP_016874096.1 7 - 8 622 113
XM_017018608.1 XP_016874097.1 9 - 10 1019 113
XM_017018609.1 XP_016874098.1 9 - 10 1310 113
XM_017018610.1 XP_016874099.1 8 - 9 1194 113
XM_017018611.1 XP_016874100.1 9 - 10 1245 113
XM_017018612.1 XP_016874101.1 10 - 11 1516 113
XM_017018613.1 XP_016874102.1 9 - 10 1310 113
XM_017018614.1 XP_016874103.1 10 - 11 1301 113
XM_017018615.1 XP_016874104.1 9 - 10 1400 113
XM_017018616.1 XP_016874105.1 8 - 9 1194 113
XM_017018617.1 XP_016874106.1 10 - 11 1399 113
XM_017018618.1 XP_016874107.1 10 - 11 1505 113
XM_017018619.1 XP_016874108.1 9 - 10 1389 113
XM_017018620.1 XP_016874109.1 9 - 10 1449 113
XM_017018621.1 XP_016874110.1 8 - 9 1333 113
XM_017018622.1 XP_016874111.1 9 - 10 1401 113
GenBank mRNA AB014552.1 - 8 - 9 903 113
AK023867.1 - 9 - 10 1012 113
AK223603.1 - 8 - 9 899 113
AK289641.1 - 9 - 10 826 113
AK293109.1 - 9 - 10 978 113
AK294110.1 - 7 - 8 854 113
BC001331.1 - 9 - 10 708 113
BC002378.2 - 9 - 10 978 113
BC006191.2 - 9 - 10 1020 113

View Full Product Details