Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278194.1 | NP_001265123.1 | 1 - 2 | 167 | 102 |
NM_198334.2 | NP_938148.1 | 1 - 2 | 167 | 102 | |
NM_198335.3 | NP_938149.2 | 1 - 2 | 167 | 102 | |
XM_017017414.1 | XP_016872903.1 | 1 - 2 | 61 | 102 | |
GenBank mRNA | BG720650.1 | - | 1 - 2 | 112 | 102 |
DC400833.1 | - | 1 - 2 | 167 | 102 | |
EU794613.1 | - | 1 - 2 | 73 | 102 |