Assay Design



Gene Transcripts

Gene Symbol
MYF5
Entrez Gene ID
4617
Gene Name
myogenic factor 5
Gene Aliases
bHLHc2
Chromosome Location
Chr.12: 80716929 - 80719668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005593.2 NP_005584.2 2 - 3 710 114
GenBank mRNA BC069373.1 - 2 - 3 617 114
BT019399.1 - 2 - 3 575 114
X14894.1 - 2 - 3 617 114

View Full Product Details