Assay Design



Gene Transcripts

Gene Symbol
VIP
Entrez Gene ID
7432
Gene Name
vasoactive intestinal peptide
Gene Aliases
PHM27
Chromosome Location
Chr.6: 152747894 - 152759765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003381.3 NP_003372.1 4 - 5 500 114
XM_006715562.3 XP_006715625.1 4 - 5 3403 114
GenBank mRNA AK313950.1 - 4 - 5 500 114
DA914915.1 - 4 - 5 500 114
M36634.1 - 4 - 5 393 114
M54930.1 - 2 - 3 180 114

View Full Product Details