Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003381.3 | NP_003372.1 | 6 - 7 | 731 | 157 |
NM_194435.2 | NP_919416.1 | 6 - 7 | 728 | 157 | |
XM_005267135.3 | XP_005267192.1 | 6 - 7 | 677 | 157 | |
XM_006715562.3 | XP_006715625.1 | 6 - 7 | 3631 | 157 | |
GenBank mRNA | AK291959.1 | - | 6 - 7 | 677 | 157 |
BC009794.1 | - | 6 - 7 | 733 | 157 | |
M36634.1 | - | 6 - 7 | 624 | 157 |