Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305581.1 | NP_001292510.1 | 5 - 6 | 597 | 91 |
NM_032024.4 | NP_114413.1 | 4 - 5 | 649 | 91 | |
NR_131178.1 | 6 - 7 | 872 | 91 | ||
XM_011540256.2 | XP_011538558.1 | 5 - 6 | 591 | 91 | |
XM_017016759.1 | XP_016872248.1 | 5 - 6 | 591 | 91 | |
XM_017016760.1 | XP_016872249.1 | 5 - 6 | 591 | 91 | |
XM_017016761.1 | XP_016872250.1 | 5 - 6 | 591 | 91 | |
GenBank mRNA | AF267860.1 | - | 4 - 5 | 650 | 91 |
BX105111.1 | - | 4 - 5 | 92 | ||
CR933639.1 | - | 6 - 7 | 872 | 91 |