Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305581.1 | NP_001292510.1 | 6 - 7 | 679 | 68 |
NM_032024.4 | NP_114413.1 | 5 - 6 | 731 | 68 | |
NR_131178.1 | 7 - 8 | 954 | 68 | ||
GenBank mRNA | AF267860.1 | - | 5 - 6 | 732 | 68 |
AK125328.1 | - | 2 - 3 | 962 | 68 | |
BX105111.1 | - | 5 - 6 | 573 | 68 | |
CR933639.1 | - | 7 - 8 | 954 | 68 |