Assay Design



Gene Transcripts

Gene Symbol
ZCCHC24
Entrez Gene ID
219654
Gene Name
zinc finger CCHC-type containing 24
Gene Aliases
C10orf56, Z3CXXC8
Chromosome Location
Chr.10: 79382325 - 79445627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153367.3 NP_699198.2 1 - 2 432 75
GenBank mRNA AK075279.1 - 1 - 2 284 75
AK308876.1 - 1 - 2 221 75
BC028617.1 - 1 - 2 434 75
BC039249.1 - 1 - 2 427 75
BC053564.1 - 1 - 2 331 75

View Full Product Details