Assay Design



Gene Transcripts

Gene Symbol
WBP11
Entrez Gene ID
51729
Gene Name
WW domain binding protein 11
Gene Aliases
NPWBP, PPP1R165, SIPP1, WBP-11
Chromosome Location
Chr.12: 14786478 - 14803467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016312.2 NP_057396.1 10 - 11 1475 76
GenBank mRNA AB029309.1 - 10 - 11 1457 76
AF118023.1 - 10 - 11 1393 76
BC001621.1 - 10 - 11 1475 76
BC016441.2 - 3 - 4 593 76
BC023532.2 - 10 - 11 1468 76

View Full Product Details