Assay Design



Gene Transcripts

Gene Symbol
WBP11
Entrez Gene ID
51729
Gene Name
WW domain binding protein 11
Gene Aliases
NPWBP, PPP1R165, SIPP1, WBP-11
Chromosome Location
Chr.12: 14786478 - 14803467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016312.2 NP_057396.1 2 - 3 227 115
GenBank mRNA AB029309.1 - 2 - 3 209 115
AF118023.1 - 2 - 3 145 115
AK297410.1 - 2 - 3 251 115
AK302244.1 - 2 - 3 265 115
BC001621.1 - 2 - 3 227 115
BC023532.2 - 2 - 3 220 115
BX647347.1 - 2 - 3 236 115

View Full Product Details