Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016312.2 | NP_057396.1 | 5 - 6 | 548 | 75 |
GenBank mRNA | AB029309.1 | - | 5 - 6 | 530 | 75 |
AF118023.1 | - | 5 - 6 | 466 | 75 | |
AK297410.1 | - | 5 - 6 | 572 | 75 | |
AK302244.1 | - | 5 - 6 | 586 | 75 | |
BC001621.1 | - | 5 - 6 | 548 | 75 | |
BC023532.2 | - | 5 - 6 | 541 | 75 |