Assay Design



Gene Transcripts

Gene Symbol
WBP11
Entrez Gene ID
51729
Gene Name
WW domain binding protein 11
Gene Aliases
NPWBP, PPP1R165, SIPP1, WBP-11
Chromosome Location
Chr.12: 14786478 - 14803467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016312.2 NP_057396.1 9 - 10 1180 136
GenBank mRNA AB029309.1 - 9 - 10 1162 136
AF118023.1 - 9 - 10 1098 136
AK297410.1 - 9 - 10 1204 136
AK302244.1 - 9 - 10 1218 136
BC001621.1 - 9 - 10 1180 136
BC016441.2 - 2 - 3 298 136
BC023532.2 - 9 - 10 1173 136
BX647347.1 - 8 - 9 924 136

View Full Product Details