Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001248011.1 | NP_001234940.1 | 4 - 5 | 873 | 112 |
NM_024047.4 | NP_076952.1 | 5 - 6 | 969 | 112 | |
NM_198038.2 | NP_932155.1 | 5 - 6 | 611 | 112 | |
XM_011532032.2 | XP_011530334.1 | 5 - 6 | 3148 | 112 | |
GenBank mRNA | AF265341.1 | - | 5 - 6 | 610 | 112 |
AF273028.1 | - | 5 - 6 | 933 | 112 | |
AK027295.1 | - | 5 - 6 | 959 | 112 | |
AK074845.1 | - | 5 - 6 | 611 | 112 | |
AK075408.1 | - | 5 - 6 | 889 | 112 | |
AK308028.1 | - | 4 - 5 | 640 | 112 | |
AY026252.1 | - | 5 - 6 | 964 | 112 | |
AY359123.1 | - | 5 - 6 | 663 | 112 | |
BC000542.2 | - | 5 - 6 | 729 | 112 | |
BI552154.1 | - | - | 611 | 113 |