Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001248011.1 | NP_001234940.1 | 5 - 6 | 1015 | 121 |
NM_024047.4 | NP_076952.1 | 6 - 7 | 1111 | 121 | |
NM_198038.2 | NP_932155.1 | 6 - 7 | 753 | 121 | |
XM_011532032.2 | XP_011530334.1 | 6 - 7 | 3290 | 121 | |
GenBank mRNA | AF265341.1 | - | 6 - 7 | 752 | 121 |
AF273028.1 | - | 6 - 7 | 1075 | 121 | |
AK074845.1 | - | 6 - 7 | 753 | 121 | |
AK075408.1 | - | 6 - 7 | 1031 | 121 | |
AK308028.1 | - | 5 - 6 | 782 | 120 | |
AY026252.1 | - | 6 - 7 | 1106 | 121 | |
AY359123.1 | - | 6 - 7 | 805 | 121 | |
BC000542.2 | - | 6 - 7 | 871 | 121 | |
BQ446599.1 | - | 2 - 3 | 121 |