Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277081.1 | NP_001264010.1 | 4 - 5 | 646 | 77 |
NM_017509.3 | NP_059979.2 | 4 - 5 | 649 | 77 | |
NR_102274.1 | 4 - 5 | 528 | 77 | ||
XM_006723265.3 | XP_006723328.1 | 5 - 6 | 1450 | 77 | |
XM_011527085.2 | XP_011525387.1 | 5 - 6 | 1446 | 77 | |
GenBank mRNA | AF303046.1 | - | 4 - 5 | 648 | 77 |
AY373373.1 | - | 3 - 4 | 738 | 77 | |
BC069480.1 | - | 4 - 5 | 646 | 77 | |
BC069507.1 | - | 4 - 5 | 649 | 77 | |
BC069518.1 | - | 4 - 5 | 646 | 77 | |
BC126137.1 | - | 4 - 5 | 639 | 77 | |
BC144045.1 | - | 4 - 5 | 518 | 77 | |
BC144046.1 | - | 4 - 5 | 636 | 77 |