Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030785.3 | NP_110412.1 | 2 - 3 | 1055 | 70 |
XM_011527351.1 | XP_011525653.1 | 2 - 3 | 1031 | 70 | |
GenBank mRNA | AK093068.1 | - | 1 - 2 | 325 | 70 |
AK098837.1 | - | 2 - 3 | 1000 | 70 | |
AK223347.1 | - | 2 - 3 | 1000 | 70 | |
AL136761.1 | - | 2 - 3 | 1027 | 70 | |
BC057785.1 | - | 2 - 3 | 883 | 70 | |
BC136351.1 | - | 2 - 3 | 936 | 70 | |
BC136352.1 | - | 2 - 3 | 936 | 70 |