Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005242.2 | NP_001005242.2 | 3 - 4 | 1153 | 73 |
NM_004572.3 | NP_004563.2 | 3 - 4 | 1153 | 73 | |
GenBank mRNA | AU125826.1 | - | 1 - 2 | 389 | 73 |
BC094762.1 | - | 3 - 4 | 1076 | 73 | |
BC126199.1 | - | 3 - 4 | 1082 | 73 | |
BC143966.1 | - | 3 - 4 | 1082 | 73 | |
EU520483.1 | - | 3 - 4 | 1038 | 73 | |
EU520484.1 | - | 3 - 4 | 1038 | 73 | |
EU520485.1 | - | 3 - 4 | 1036 | 73 | |
EU520486.1 | - | 3 - 4 | 1038 | 73 | |
X97675.1 | - | 3 - 4 | 1063 | 73 |