Assay Design



Gene Transcripts

Gene Symbol
TSNARE1
Entrez Gene ID
203062
Gene Name
t-SNARE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 142212080 - 142403291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145003.4 NP_659440.2 2 - 3 271 140
XM_011516912.1 XP_011515214.1 1 - 2 125 140
XM_011516913.2 XP_011515215.1 2 - 3 313 140
XM_011516914.1 XP_011515216.1 2 - 3 224 140
XM_011516915.2 XP_011515217.1 3 - 4 310 140
XM_011516916.1 XP_011515218.1 2 - 3 313 140
XM_011516917.2 XP_011515219.1 2 - 3 150 140
XM_011516919.1 XP_011515221.1 1 - 2 125 140
XM_011516921.1 XP_011515223.1 1 - 2 125 140
XM_011516922.1 XP_011515224.1 1 - 2 125 140
XM_011516924.1 XP_011515226.1 1 - 2 125 140
XM_017013175.1 XP_016868664.1 1 - 2 125 140
XM_017013176.1 XP_016868665.1 1 - 2 125 140
XM_017013177.1 XP_016868666.1 1 - 2 125 140
XM_017013178.1 XP_016868667.1 1 - 2 125 140
XM_017013179.1 XP_016868668.1 1 - 2 125 140
XM_017013180.1 XP_016868669.1 1 - 2 125 140
XM_017013181.1 XP_016868670.1 1 - 2 125 140
GenBank mRNA AK055726.1 - 2 - 3 200 140
BC113556.1 - 1 - 2 125 140
BC126343.1 - 1 - 2 125 140
BC143684.1 - 1 - 2 125 140
BC143686.1 - 1 - 2 125 140
DA229159.1 - 2 - 3 204 140
HY009343.1 - 2 - 3 319 140

View Full Product Details