Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022569.2 | NP_072091.1 | 13 - 14 | 3184 | 101 |
XM_017008545.1 | XP_016864034.1 | 12 - 13 | 2182 | 101 | |
XM_017008546.1 | XP_016864035.1 | 11 - 12 | 1488 | 101 | |
GenBank mRNA | AB036429.1 | - | 13 - 14 | 3184 | 101 |
AK289670.1 | - | 13 - 14 | 3162 | 101 | |
BC113075.2 | - | 12 - 13 | 1871 | 101 |