Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002943.3 | NP_002934.1 | 10 - 11 | 1640 | 132 |
NM_134260.2 | NP_599022.1 | 11 - 12 | 1664 | 132 | |
NM_134261.2 | NP_599023.1 | 10 - 11 | 1494 | 132 | |
NM_134262.2 | NP_599024.1 | 9 - 10 | 1335 | 132 | |
XM_005254584.4 | XP_005254641.1 | 10 - 11 | 5955 | 132 | |
XM_011521874.1 | XP_011520176.1 | 9 - 10 | 1532 | 132 | |
XM_011521875.1 | XP_011520177.1 | 10 - 11 | 1364 | 132 | |
XM_011521877.2 | XP_011520179.1 | 9 - 10 | 1567 | 132 | |
XM_011521878.1 | XP_011520180.1 | 11 - 12 | 1406 | 132 | |
XM_011521879.2 | XP_011520181.1 | 7 - 8 | 1292 | 132 | |
XM_017022466.1 | XP_016877955.1 | 9 - 10 | 3356 | 132 | |
XM_017022467.1 | XP_016877956.1 | 9 - 10 | 3029 | 132 | |
GenBank mRNA | AB307694.1 | - | 10 - 11 | 1409 | 132 |
BC008831.2 | - | 10 - 11 | 1417 | 132 | |
BC100987.2 | - | 9 - 10 | 1285 | 132 | |
BC100988.1 | - | 9 - 10 | 1285 | 132 | |
BC100989.1 | - | 9 - 10 | 1285 | 132 | |
BC100990.2 | - | 9 - 10 | 1285 | 132 | |
HQ692818.1 | - | 9 - 10 | 1244 | 132 | |
HQ709174.1 | - | 9 - 10 | 1241 | 132 | |
L14611.1 | - | 9 - 10 | 132 | ||
U04897.1 | - | 10 - 11 | 1510 | 132 | |
U04898.1 | - | 11 - 12 | 1597 | 132 | |
U04899.1 | - | 10 - 11 | 1562 | 132 |