Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002943.3 | NP_002934.1 | 3 - 4 | 512 | 85 |
NM_134260.2 | NP_599022.1 | 4 - 5 | 536 | 85 | |
NM_134261.2 | NP_599023.1 | 3 - 4 | 366 | 85 | |
NM_134262.2 | NP_599024.1 | 2 - 3 | 207 | 85 | |
XM_005254584.4 | XP_005254641.1 | 3 - 4 | 4827 | 85 | |
XM_011521874.1 | XP_011520176.1 | 2 - 3 | 404 | 85 | |
XM_011521875.1 | XP_011520177.1 | 3 - 4 | 236 | 85 | |
XM_011521877.2 | XP_011520179.1 | 2 - 3 | 439 | 85 | |
XM_011521878.1 | XP_011520180.1 | 4 - 5 | 278 | 85 | |
XM_017022466.1 | XP_016877955.1 | 2 - 3 | 2228 | 85 | |
XM_017022467.1 | XP_016877956.1 | 2 - 3 | 1901 | 85 | |
GenBank mRNA | AB307694.1 | - | 3 - 4 | 281 | 85 |
AL532048.3 | - | 2 - 3 | 207 | 85 | |
BC008831.2 | - | 3 - 4 | 289 | 85 | |
BC100987.2 | - | 2 - 3 | 157 | 85 | |
BC100988.1 | - | 2 - 3 | 157 | 85 | |
BC100989.1 | - | 2 - 3 | 157 | 85 | |
BC100990.2 | - | 2 - 3 | 157 | 85 | |
BG705319.1 | - | 2 - 3 | 387 | 85 | |
DB482516.1 | - | 2 - 3 | 379 | 85 | |
HQ692818.1 | - | 2 - 3 | 116 | 85 | |
HQ709174.1 | - | 2 - 3 | 116 | 85 | |
L14611.1 | - | 2 - 3 | 137 | 85 | |
U04897.1 | - | 3 - 4 | 382 | 85 | |
U04898.1 | - | 4 - 5 | 469 | 85 | |
U04899.1 | - | 3 - 4 | 434 | 85 |