Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002943.3 | NP_002934.1 | 7 - 8 | 1308 | 100 |
NM_134260.2 | NP_599022.1 | 8 - 9 | 1332 | 100 | |
NM_134261.2 | NP_599023.1 | 7 - 8 | 1162 | 100 | |
NM_134262.2 | NP_599024.1 | 6 - 7 | 1003 | 100 | |
XM_005254584.4 | XP_005254641.1 | 7 - 8 | 5623 | 100 | |
XM_011521874.1 | XP_011520176.1 | 6 - 7 | 1200 | 100 | |
XM_011521875.1 | XP_011520177.1 | 7 - 8 | 1032 | 100 | |
XM_011521877.2 | XP_011520179.1 | 6 - 7 | 1235 | 100 | |
XM_011521878.1 | XP_011520180.1 | 8 - 9 | 1074 | 100 | |
XM_011521879.2 | XP_011520181.1 | 4 - 5 | 960 | 100 | |
XM_017022466.1 | XP_016877955.1 | 6 - 7 | 3024 | 100 | |
XM_017022467.1 | XP_016877956.1 | 6 - 7 | 2697 | 100 | |
GenBank mRNA | AB307694.1 | - | 7 - 8 | 1077 | 100 |
BC008831.2 | - | 7 - 8 | 1085 | 100 | |
BC100987.2 | - | 6 - 7 | 953 | 100 | |
BC100988.1 | - | 6 - 7 | 953 | 100 | |
BC100989.1 | - | 6 - 7 | 953 | 100 | |
BC100990.2 | - | 6 - 7 | 953 | 100 | |
HQ692818.1 | - | 6 - 7 | 912 | 100 | |
HQ709174.1 | - | 6 - 7 | 909 | 100 | |
U04897.1 | - | 7 - 8 | 1178 | 100 | |
U04898.1 | - | 8 - 9 | 1265 | 100 | |
U04899.1 | - | 7 - 8 | 1230 | 100 |