Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012175.3 | NP_036307.2 | 6 - 7 | 747 | 96 |
NM_033406.2 | NP_208385.1 | 6 - 7 | 747 | 96 | |
XM_011519980.1 | XP_011518282.1 | 6 - 7 | 753 | 96 | |
XM_011519981.2 | XP_011518283.1 | 6 - 7 | 753 | 96 | |
XM_011519982.1 | XP_011518284.1 | 7 - 8 | 807 | 96 | |
XM_017017533.1 | XP_016873022.1 | 7 - 8 | 807 | 96 | |
GenBank mRNA | AB209039.1 | - | 5 - 6 | 4132 | 96 |
AF176702.1 | - | 6 - 7 | 704 | 96 | |
AK001943.1 | - | 6 - 7 | 738 | 96 | |
AK128454.1 | - | 6 - 7 | 725 | 96 | |
AK304866.1 | - | 5 - 6 | 537 | 96 | |
AL832826.1 | - | 5 - 6 | 1419 | 96 | |
BC046110.1 | - | 6 - 7 | 733 | 96 |