Assay Design



Gene Transcripts

Gene Symbol
LAMP1
Entrez Gene ID
3916
Gene Name
lysosomal associated membrane protein 1
Gene Aliases
CD107a, LAMPA, LGP120
Chromosome Location
Chr.13: 113297154 - 113323431 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005561.3 NP_005552.3 7 - 8 1227 137
XM_011537494.1 XP_011535796.1 7 - 8 904 137
GenBank mRNA AB209319.1 - 5 - 6 3374 137
AK092398.1 - 8 - 9 1104 137
AK301584.1 - 6 - 7 838 137
BC006345.2 - 5 - 6 621 137
BC007845.2 - 4 - 5 439 137
BC021288.2 - 7 - 8 1002 137
BC025335.1 - 7 - 8 1140 137
BC093044.1 - 7 - 8 1095 137
J03263.1 - 6 - 7 874 137
J04182.1 - 7 - 8 1133 137

View Full Product Details