Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005561.3 | NP_005552.3 | 8 - 9 | 1399 | 121 |
XM_011537494.1 | XP_011535796.1 | 8 - 9 | 1076 | 121 | |
GenBank mRNA | AK092398.1 | - | 9 - 10 | 1276 | 121 |
AK301584.1 | - | 7 - 8 | 1010 | 121 | |
BC006345.2 | - | 6 - 7 | 793 | 121 | |
BC007845.2 | - | 5 - 6 | 611 | 121 | |
BC021288.2 | - | 8 - 9 | 1174 | 121 | |
BC025335.1 | - | 8 - 9 | 1312 | 121 | |
BC093044.1 | - | 8 - 9 | 1267 | 121 | |
J03263.1 | - | 7 - 8 | 1046 | 121 | |
J04182.1 | - | 8 - 9 | 1305 | 121 |