Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201564.1 | NP_001188493.1 | 12 - 13 | 1248 | 139 |
NM_002669.3 | NP_002660.1 | 12 - 13 | 1275 | 139 | |
GenBank mRNA | AF044333.1 | - | 11 - 12 | 1150 | 139 |
AK002108.1 | - | 12 - 13 | 1250 | 139 | |
BC020786.1 | - | 12 - 13 | 1212 | 139 | |
BC106004.1 | - | 12 - 13 | 1213 | 139 |