Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201564.1 | NP_001188493.1 | 2 - 3 | 236 | 74 |
NM_002669.3 | NP_002660.1 | 2 - 3 | 236 | 74 | |
GenBank mRNA | AF044333.1 | - | 1 - 2 | 111 | 74 |
AK002108.1 | - | 2 - 3 | 211 | 74 | |
AK304574.1 | - | 2 - 3 | 195 | 74 | |
AK307997.1 | - | 2 - 3 | 183 | 74 | |
BC020786.1 | - | 2 - 3 | 200 | 74 | |
BC106004.1 | - | 2 - 3 | 174 | 74 | |
BP314297.1 | - | 2 - 3 | 236 | 74 |