Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002669.3 | NP_002660.1 | 3 - 4 | 386 | 104 |
GenBank mRNA | AF044333.1 | - | 2 - 3 | 261 | 104 |
AK002108.1 | - | 3 - 4 | 361 | 104 | |
AK304574.1 | - | 3 - 4 | 345 | 104 | |
AK307997.1 | - | 3 - 4 | 333 | 104 | |
BC106004.1 | - | 3 - 4 | 324 | 104 | |
BP314297.1 | - | 3 - 4 | 386 | 104 |