Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201564.1 | NP_001188493.1 | 8 - 9 | 789 | 96 |
NM_002669.3 | NP_002660.1 | 8 - 9 | 816 | 96 | |
GenBank mRNA | AF044333.1 | - | 7 - 8 | 691 | 96 |
AK002108.1 | - | 8 - 9 | 791 | 96 | |
AK304574.1 | - | 7 - 8 | 684 | 96 | |
BC020786.1 | - | 8 - 9 | 753 | 96 | |
BC106004.1 | - | 8 - 9 | 754 | 96 |