Assay Design



Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
56942
Gene Name
C-X9-C motif containing 2
Gene Aliases
2310061C15Rik, C16orf61, DC13
Chromosome Location
Chr.16: 80975800 - 81007046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017023461.1 XP_016878950.1 2 - 3 413 154
XM_017023462.1 XP_016878951.1 2 - 3 428 154

View Full Product Details