Assay Design



Gene Transcripts

Gene Symbol
SERPINB11
Entrez Gene ID
89778
Gene Name
serpin family B member 11 (gene/pseudogene)
Gene Aliases
EPIPIN, SERPIN11
Chromosome Location
Chr.18: 63702304 - 63723893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291278.1 NP_001278207.1 2 - 3 230 110
NM_001291279.1 NP_001278208.1 2 - 3 230 110
NM_080475.3 NP_536723.2 3 - 4 412 110
XM_011526251.1 XP_011524553.1 2 - 3 453 110
GenBank mRNA AF419953.1 - 1 - 2 167 110
AF419954.1 - 1 - 2 167 110
AF419955.1 - 1 - 2 167 110
AK292775.1 - 2 - 3 229 110
AK309537.1 - 3 - 4 315 110
AK309970.1 - 2 - 3 229 110
AY739645.1 - 3 - 4 411 110
AY792323.1 - 1 - 2 167 110
AY792324.1 - 1 - 2 167 110
AY792325.1 - 1 - 2 167 110
BC069596.1 - 1 - 2 167 110
BC130370.1 - 1 - 2 193 110
BC130372.1 - 1 - 2 193 110
DC365081.1 - 2 - 3 229 110

View Full Product Details