Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 12 - 13 | 1343 | 101 |
NM_001286458.1 | NP_001273387.1 | 13 - 14 | 1518 | 101 | |
NM_001321858.1 | NP_001308787.1 | 12 - 13 | 1269 | 101 | |
NM_003470.2 | NP_003461.2 | 12 - 13 | 1474 | 101 | |
NR_135826.1 | 12 - 13 | 1297 | 101 | ||
XM_017023652.1 | XP_016879141.1 | 12 - 13 | 1902 | 101 | |
XM_017023653.1 | XP_016879142.1 | 12 - 13 | 1338 | 101 | |
GenBank mRNA | AK302481.1 | - | 13 - 14 | 1444 | 101 |
AK302771.1 | - | 12 - 13 | 1275 | 101 | |
AK302872.1 | - | 12 - 13 | 1297 | 101 | |
AK302912.1 | - | 13 - 14 | 1462 | 101 | |
AK316441.1 | - | 12 - 13 | 1344 | 101 | |
AY376241.1 | - | 12 - 13 | 1305 | 101 | |
Z72499.1 | - | 12 - 13 | 1474 | 101 |