Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 21 - 22 | 2378 | 83 |
NM_001286458.1 | NP_001273387.1 | 22 - 23 | 2553 | 83 | |
NM_001321858.1 | NP_001308787.1 | 21 - 22 | 2304 | 83 | |
NM_003470.2 | NP_003461.2 | 21 - 22 | 2509 | 83 | |
NR_135826.1 | 21 - 22 | 2291 | 83 | ||
XM_017023652.1 | XP_016879141.1 | 21 - 22 | 2937 | 83 | |
XM_017023653.1 | XP_016879142.1 | 21 - 22 | 2373 | 83 | |
GenBank mRNA | AK302481.1 | - | 22 - 23 | 2479 | 83 |
AK302771.1 | - | 21 - 22 | 2310 | 83 | |
AK302912.1 | - | 22 - 23 | 2497 | 83 | |
AK316441.1 | - | 21 - 22 | 2379 | 83 | |
AY376241.1 | - | 21 - 22 | 2340 | 83 | |
Z72499.1 | - | 21 - 22 | 2509 | 83 |