Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 24 - 25 | 2708 | 136 |
NM_001286458.1 | NP_001273387.1 | 25 - 26 | 2883 | 136 | |
NM_001321858.1 | NP_001308787.1 | 24 - 25 | 2634 | 136 | |
NM_003470.2 | NP_003461.2 | 24 - 25 | 2839 | 136 | |
NR_135826.1 | 24 - 25 | 2621 | 136 | ||
XM_017023652.1 | XP_016879141.1 | 24 - 25 | 3267 | 136 | |
XM_017023653.1 | XP_016879142.1 | 24 - 25 | 2703 | 136 | |
GenBank mRNA | AK302481.1 | - | 25 - 26 | 2809 | 136 |
AK302771.1 | - | 24 - 25 | 2640 | 136 | |
AK302912.1 | - | 25 - 26 | 2827 | 136 | |
AK316441.1 | - | 24 - 25 | 2709 | 136 | |
AY376241.1 | - | 24 - 25 | 2670 | 136 | |
Z72499.1 | - | 24 - 25 | 2839 | 136 |