Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 30 - 31 | 3270 | 93 |
NM_001286458.1 | NP_001273387.1 | 31 - 32 | 3445 | 93 | |
NM_001321858.1 | NP_001308787.1 | 30 - 31 | 3196 | 93 | |
NM_003470.2 | NP_003461.2 | 30 - 31 | 3401 | 93 | |
NR_135826.1 | 30 - 31 | 3183 | 93 | ||
GenBank mRNA | AK302771.1 | - | 30 - 31 | 3202 | 93 |
AK302912.1 | - | 31 - 32 | 3389 | 93 | |
AK316441.1 | - | 30 - 31 | 3271 | 93 | |
AY376241.1 | - | 30 - 31 | 3232 | 93 | |
Z72499.1 | - | 30 - 31 | 3401 | 93 |